• USA Home
  • Search Results

  • 5iodoacetamidofluorescein515256336854711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-(Iodoacetamido)fluorescein

1 Products
Synonym:   5-IAF
Image of 5-<WBR>(Iodoacetamido)<WBR>fluorescein
CAS Number:
Empirical Formula (Hill Notation):
 C22H14INO6
Molecular Weight:
 515.25
 Beilstein/REAXYS Number:
 6543244
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Fluorescence Test
  Application(s)
Show More