• USA Home
  • Search Results

  • 5methoxyindole14717100694611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Methoxyindole

1 Products
Image of 5-Methoxyindole
CAS Number:
Empirical Formula (Hill Notation):
 C9H9NO
Molecular Weight:
 147.17
 Beilstein/REAXYS Number:
 116722
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
M14900 5-Methoxyindole, 99%,