• USA Home
  • Search Results

  • 5methoxyindole3carboxaldehyde175181060119111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Methoxyindole-3-carboxaldehyde

1 Products
Synonym:   3-Formyl-5-methoxyindole, NSC 521754
Image of 5-<WBR>Methoxyindole-<WBR>3-<WBR>carboxaldehyde
CAS Number:
Empirical Formula (Hill Notation):
 C10H9NO2
Molecular Weight:
 175.18
 Beilstein/REAXYS Number:
 132769
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More