• USA Home
  • Search Results

  • 5methoxytryptamine1902460807111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Methoxytryptamine

2 Products
Synonym:   2-(5-Methoxyindol-3-yl)ethylamine, 3-(2-Aminoethyl)-5-methoxyindole, 5-Methoxyindole-3-ethanamine, 5-MOT, Deacetylmelatonin, NSC 56422
Image of 5-Methoxytryptamine
CAS Number:
Empirical Formula (Hill Notation):
 C11H14N2O
Molecular Weight:
 190.24
 Beilstein/REAXYS Number:
 145587
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More