• USA Home
  • Search Results

  • 5methyl2hexanol1162062759811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Methyl-2-hexanol

1 Products
Image of 5-Methyl-2-hexanol
CAS Number:
Linear Formula:
 (CH3)2CHCH2CH2CHOHCH3
Molecular Weight:
 116.20
 Beilstein/REAXYS Number:
 1731619
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
189731 5-Methyl-2-hexanol, 98%,