• USA Home
  • Search Results

  • 5methyl2thiophenecarboxylicacid14218191879211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Methyl-2-thiophenecarboxylic acid

1 Products
Image of 5-<WBR>Methyl-<WBR>2-<WBR>thiophenecarboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H6O2S
Molecular Weight:
 142.18
 Beilstein/REAXYS Number:
 113857
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More