• USA Home
  • Search Results

  • 5methyluridine25823146310111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Methyluridine

1 Products
Synonym:   Ribothymidine
Image of 5-Methyluridine
CAS Number:
Empirical Formula (Hill Notation):
 C10H14N2O6
Molecular Weight:
 258.23
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
535893 5-Methyluridine, 97%,