• USA Home
  • Search Results

  • 5oxo5phenylvalericacid19221150105911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Oxo-5-phenylvaleric acid

1 Products
Synonym:   4-Benzoylbutyric acid
Image of 5-<WBR>Oxo-<WBR>5-<WBR>phenylvaleric acid
CAS Number:
Linear Formula:
 C6H5CO(CH2)3CO2H
Molecular Weight:
 192.21
 Beilstein/REAXYS Number:
 641947
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More