• USA Home
  • Search Results

  • 610dimethyl59undecadien2one1943168967811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6,10-Dimethyl-5,9-undecadien-2-one

1 Products
Synonym:   6,10-Dimethyl-5,9-undecadien-2-one, mixture of isomers, Geranylacetone
Image of 6,10-<WBR>Dimethyl-<WBR>5,9-<WBR>undecadien-<WBR>2-<WBR>one
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2COCH3
Molecular Weight:
 194.31
 Beilstein/REAXYS Number:
 1722277
 Council of Europe no.:
 11088
 EC Number:
 FEMA Number:
 3542
 Flavis number:
 7.123
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
Show More