• USA Home
  • Search Results

  • 69diamino2ethoxyacridinedllactatemonohydrate36139640223911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6,9-Diamino-2-ethoxyacridine-DL-lactate monohydrate

1 Products
Synonym:   Acrinol, Ethacridine lactate salt, Ethodin
Image of 6,9-<WBR>Diamino-<WBR>2-<WBR>ethoxyacridine-<SC>DL</SC>-lactate monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C15H15N3O · C3H6O3 · H2O
Molecular Weight:
 361.39
 Beilstein/REAXYS Number:
 3642805
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More