• USA Home
  • Search Results

  • 6aminocaproicacid131176032211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Aminocaproic acid

2 Products
Synonym:   ε-Aminocaproic acid, 6-Aminohexanoic acid, EACA
Image of 6-Aminocaproic acid
CAS Number:
Linear Formula:
 H2N(CH2)5CO2H
Molecular Weight:
 131.17
 Beilstein/REAXYS Number:
 906872
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Biological Source
  Physical Form
  Application(s)
  Solubility
Show More