• USA Home
  • Search Results

  • 6aminopyridine3carboxylicacid13812316749511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Aminopyridine-3-carboxylic acid

1 Products
Synonym:   6-Aminonicotinic acid
Image of 6-<WBR>Aminopyridine-<WBR>3-<WBR>carboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H6N2O2
Molecular Weight:
 138.12
 Beilstein/REAXYS Number:
 115992
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More