• USA Home
  • Search Results

  • 6bromo1hexanol18107428655911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Bromo-1-hexanol

1 Products
Synonym:   Hexamethylene bromohydrin
Image of 6-Bromo-1-hexanol
CAS Number:
Linear Formula:
 Br(CH2)6OH
Molecular Weight:
 181.07
 Beilstein/REAXYS Number:
 1732415
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
186481 6-Bromo-1-hexanol, 97%,