• USA Home
  • Search Results

  • 6bromo2naphthol223071523191111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Bromo-2-naphthol

1 Products
Image of 6-Bromo-2-naphthol
CAS Number:
Linear Formula:
 BrC10H6OH
Molecular Weight:
 223.07
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
B73406 6-Bromo-2-naphthol, 97%,