• USA Home
  • Search Results

  • 6bromohexanoylchloride213502280937611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Bromohexanoyl chloride

1 Products
Synonym:   6-Bromocaproyl chloride, 6-Bromohexanoic acid chloride
Image of 6-Bromohexanoyl chloride
CAS Number:
Linear Formula:
 Br(CH2)5COCl
Molecular Weight:
 213.50
 Beilstein/REAXYS Number:
 1749739
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More