• USA Home
  • Search Results

  • 6chloro24diamino135triazine13c314853121685033711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Chloro-2,4-diamino-1,3,5-triazine-13C3

2 Products
Image of 6-<WBR>Chloro-<WBR>2,4-<WBR>diamino-<WBR>1,3,5-<WBR>triazine-<SUP>13</SUP>C<SUB>3</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 13C3H4ClN5
Molecular Weight:
 148.53
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More