• USA Home
  • Search Results

  • 6chloro3methyluracil16056431856311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Chloro-3-methyluracil

1 Products
Synonym:   6-Chloro-3-methylpyrimidine-2,4(1H,3H)-dione
Image of 6-<WBR>Chloro-<WBR>3-<WBR>methyluracil
CAS Number:
Empirical Formula (Hill Notation):
 C5H5ClN2O2
Molecular Weight:
 160.56
 Beilstein/REAXYS Number:
 511456
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More