• USA Home
  • Search Results

  • 6chloropyridine3carboxylicacid15755532623811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Chloropyridine-3-carboxylic acid

2 Products
Synonym:   6-Chloronicotinic acid
Image of 6-<WBR>Chloropyridine-<WBR>3-<WBR>carboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H4ClNO2
Molecular Weight:
 157.55
 Beilstein/REAXYS Number:
 115993
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More