• USA Home
  • Search Results

  • 6fluorescein56carboxamidohexanoicacidnhydroxysuccinimideester5865511461631811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-[Fluorescein-5(6)-carboxamido]hexanoic acid N-hydroxysuccinimide ester

1 Products
Synonym:   Fluorescein-5(6)-carboxamidocaproic acid N-succinimidyl ester
Image of 6-<WBR>[Fluorescein-<WBR>5(6)<WBR>-<WBR>carboxamido]<WBR>hexanoic acid <I>N</I>-hydroxysuccinimide ester
CAS Number:
Empirical Formula (Hill Notation):
 C31H26N2O10
Molecular Weight:
 586.55
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Fluorescence Test
  Solubility
Show More