• USA Home
  • Search Results

  • 6gingerol294392351314611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

[6]-Gingerol

3 Products
Synonym:   3-Decanone, 5-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-, (5S)-, 5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone, 6-Gingerol
Image of [6]-Gingerol
CAS Number:
Empirical Formula (Hill Notation):
 C17H26O4
Molecular Weight:
 294.39
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More