• USA Home
  • Search Results

  • 6hexachlorofluoresceinphosphoramidite105061136054755211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Hexachloro-Fluorescein Phosphoramidite

3 Products
Synonym:   6-(4,7,2′,4′,5′,7′-Hexachloro-3′,6′-dipivaloylfluoresceinyl-6-carboxamido)-hexyl-1-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Image of 6-<WBR>Hexachloro-<WBR>Fluorescein Phosphoramidite
CAS Number:
Empirical Formula (Hill Notation):
 C46H52Cl6N3O10P
Molecular Weight:
 1050.61
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Compatibility
Show More