• USA Home
  • Search Results

  • 6hydroxychlorzoxazone18556175045411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Hydroxychlorzoxazone

1 Products
Synonym:   5-Chloro-6-hydroxy-2(3H)-benzoxazolone, 5-Chloro-6-hydroxybenzoxazone
Image of 6-<WBR>Hydroxychlorzoxazone
CAS Number:
Empirical Formula (Hill Notation):
 C7H4ClNO3
Molecular Weight:
 185.56
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More