• USA Home
  • Search Results

  • 6hydroxypyridine3carboxylicacid13911500666611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Hydroxypyridine-3-carboxylic acid

2 Products
Synonym:   2-Hydroxy-5-pyridinecarboxylic acid, 6-Hydroxynicotinic acid
Image of 6-<WBR>Hydroxypyridine-<WBR>3-<WBR>carboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H5NO3
Molecular Weight:
 139.11
 Beilstein/REAXYS Number:
 115991
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More