• USA Home
  • Search Results

  • 6maleimidohexanoicacidnhydroxysuccinimideester308295575063511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Maleimidohexanoic acid N-hydroxysuccinimide ester

3 Products
Synonym:   6-Maleimidocaproic acid N-succinimidyl ester, N-(ε-Maleimidocaproyloxy)succinimide, N-Succinimidyl 6-maleimidocaproate, EMCS
Image of 6-Maleimidohexanoic acid <I>N</I>-hydroxysuccinimide ester
CAS Number:
Empirical Formula (Hill Notation):
 C14H16N2O6
Molecular Weight:
 308.29
 Beilstein/REAXYS Number:
 1499815
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More