• USA Home
  • Search Results

  • 6mercaptopurinemonohydrate17019611276111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Mercaptopurine monohydrate

1 Products
Synonym:   6-Purinethiol, 6-Thiohypoxanthine
Image of 6-Mercaptopurine monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C5H4N4S · H2O
Molecular Weight:
 170.19
 Beilstein/REAXYS Number:
 4012091
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More