• USA Home
  • Search Results

  • 6methoxyindole14717318913711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Methoxyindole

1 Products
Synonym:   NSC 92517
Image of 6-Methoxyindole
CAS Number:
Empirical Formula (Hill Notation):
 C9H9NO
Molecular Weight:
 147.17
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
139858 6-Methoxyindole, 98%,