• USA Home
  • Search Results

  • 6methyl5hepten2one1262011093011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Methyl-5-hepten-2-one

4 Products
Image of 6-<WBR>Methyl-<WBR>5-<WBR>hepten-<WBR>2-<WBR>one
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2COCH3
Molecular Weight:
 126.20
 Beilstein/REAXYS Number:
 1741705
 Council of Europe no.:
 149c
 EC Number:
 FEMA Number:
 2707
 Flavis number:
 7.015
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Refractive Index
  Document Links
  Special Grade
  Allergen
  Application(s)
  Organoleptic property
  Agency/Method
  Assay
Show More