• USA Home
  • Search Results

  • 6nitrohexanoicacid161161026996211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

6-Nitrohexanoic acid

1 Products
Synonym:   6-Nitrocaproic acid
Image of 6-Nitrohexanoic acid
CAS Number:
Linear Formula:
 O2N(CH2)5CO2H
Molecular Weight:
 161.16
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
346268 6-Nitrohexanoic acid, 98%,