• USA Home
  • Search Results

  • 78dihydroxycoumarin1781448635111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

7,8-Dihydroxycoumarin

1 Products
Synonym:   Daphnetin, Daphnetol, Ruixiangsu
Image of 7,8-<WBR>Dihydroxycoumarin
CAS Number:
Empirical Formula (Hill Notation):
 C9H6O4
Molecular Weight:
 178.14
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More