• USA Home
  • Search Results

  • 7allyl78dihydro8oxoguanosine3393012128839911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

7-Allyl-7,8-dihydro-8-oxoguanosine

1 Products
Synonym:   Loxoribine
Image of 7-<WBR>Allyl-<WBR>7,8-<WBR>dihydro-<WBR>8-<WBR>oxoguanosine
CAS Number:
Empirical Formula (Hill Notation):
 C13H17N5O6
Molecular Weight:
 339.30
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More