• USA Home
  • Search Results

  • 7amino4methylcoumarin175182609331211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

7-Amino-4-methylcoumarin

2 Products
Synonym:   Coumarin 120
Image of 7-<WBR>Amino-<WBR>4-<WBR>methylcoumarin
CAS Number:
Empirical Formula (Hill Notation):
 C10H9NO2
Molecular Weight:
 175.18
 Beilstein/REAXYS Number:
 142231
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More