• USA Home
  • Search Results

  • 7deaza2deoxyguanosine5triphosphate5061910151508611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

7-Deaza-2′-deoxy-guanosine-5′-triphosphate

1 Products
Synonym:   −N7-dGTP, 7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt, 7-Deaza-dGTP
Image of 7-Deaza-2′-deoxy-guanosine-5′-triphosphate
CAS Number:
Empirical Formula (Hill Notation):
 C11H17N4O13P3
Molecular Weight:
 506.19
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More