• USA Home
  • Search Results

  • 7dehydrocholesterol3846443416211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

7-Dehydrocholesterol

1 Products
Synonym:   (−)-7-Dehydrocholesterol, 3β-Hydroxy-5,7-cholestadiene, 5,7-Cholestadien-3β-ol, Provitamin D3
Image of 7-Dehydrocholesterol
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O
Molecular Weight:
 384.64
 Beilstein/REAXYS Number:
 2224615
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
 eCl@ss
 39023139
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More