• USA Home
  • Search Results

  • 7hydroxyflavanone24025651536211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

7-Hydroxyflavanone

1 Products
Synonym:   7-Hydroxy-2-phenylchroman-4-one
Image of 7-Hydroxyflavanone
CAS Number:
Empirical Formula (Hill Notation):
 C15H12O3
Molecular Weight:
 240.25
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
419753 7-Hydroxyflavanone, 98%,