• USA Home
  • Search Results

  • 7methoxycoumarin4aceticacid234206293572211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

7-Methoxycoumarin-4-acetic acid

1 Products
Synonym:   7-Methoxy-2-oxo-2H-1-benzopyran-4-acetic acid
Image of 7-<WBR>Methoxycoumarin-<WBR>4-<WBR>acetic acid
CAS Number:
Empirical Formula (Hill Notation):
 C12H10O5
Molecular Weight:
 234.20
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More