• USA Home
  • Search Results

  • 8bromoadenosine34614294639611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

8-Bromoadenosine

1 Products
Synonym:   6-Amino-8-bromopurine riboside, 8-Bromoadenine-9-β-D-ribofuranoside
Image of 8-Bromoadenosine
CAS Number:
Empirical Formula (Hill Notation):
 C10H12BrN5O4
Molecular Weight:
 346.14
 Beilstein Registry Number:
 627737
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
B6272 8-Bromoadenosine, powder,