• USA Home
  • Search Results

  • 8hydroxyquinoline1451614824311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

8-Hydroxyquinoline

4 Products
Synonym:   Oxine, 8-Quinolinol
Image of 8-Hydroxyquinoline
CAS Number:
Empirical Formula (Hill Notation):
 C9H7NO
Molecular Weight:
 145.16
 EC Index Number
 205-711-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Mode of Action
  Activity Spectrum
  Assay
Show More