• USA Home
  • Search Results

  • 8prenylnaringenin340376868202011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

8-Prenylnaringenin

2 Products
Synonym:   2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-8-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one
Image of 8-Prenylnaringenin
CAS Number:
Empirical Formula (Hill Notation):
 C20H20O5
Molecular Weight:
 340.37
 Beilstein/REAXYS Number:
 5611472
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More