• USA Home
  • Search Results

  • 8quinolinol1451614824311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

8-Quinolinol

2 Products
Synonym:   8-Hydroxyquinoline, 8-Oxychinolin, Oxine
Image of 8-Quinolinol
CAS Number:
Empirical Formula (Hill Notation):
 C9H7NO
Molecular Weight:
 145.16
 Beilstein/REAXYS Number:
 114512
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Mode of Action
  Titertest
  Purity Grade
  Melting Point
  Activity Spectrum
  Special Grade
  Assay
Show More