• USA Home
  • Search Results

  • 8quinolinolhemisulfatesalt3883913431611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

8-Quinolinol hemisulfate salt

1 Products
Synonym:   8-Hydroxyquinoline hemisulfate salt
Image of 8-Quinolinol hemisulfate salt
CAS Number:
Empirical Formula (Hill Notation):
 C18H14N2O2 · H2SO4
Molecular Weight:
 388.39
 Beilstein/REAXYS Number:
 3760550
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Mode of Action
  Activity Spectrum
  Assay
Show More