• USA Home
  • Search Results

  • 910phenanthrenequinone208218411711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

9,10-Phenanthrenequinone

3 Products
Synonym:   9,10-Phenanthrenedione
Image of 9,10-<WBR>Phenanthrenequinone
CAS Number:
Empirical Formula (Hill Notation):
 C14H8O2
Molecular Weight:
 208.21
 Beilstein/REAXYS Number:
 608838
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More