• USA Home
  • Search Results

  • 9borabicyclo331nonanesolution1220228064811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

9-Borabicyclo[3.3.1]nonane solution

2 Products
Synonym:   9-BBN
Image of 9-<WBR>Borabicyclo[3.3.1]<WBR>nonane solution
CAS Number:
Empirical Formula (Hill Notation):
 C8H15B
Molecular Weight:
 122.02
 Beilstein/REAXYS Number:
 605509
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More