• USA Home
  • Search Results

  • 9bromo1nonanol223155536280611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

9-Bromo-1-nonanol

1 Products
Synonym:   Nonamethylene bromohydrin
Image of 9-Bromo-1-nonanol
CAS Number:
Linear Formula:
 Br(CH2)9OH
Molecular Weight:
 223.15
 Beilstein/REAXYS Number:
 1737525
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
448729 9-Bromo-1-nonanol, 95%,