• USA Home
  • Search Results

  • 9chloro9phenylxanthene292764250603611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

9-Chloro-9-phenylxanthene

1 Products
Synonym:   Pixyl chloride
Image of 9-<WBR>Chloro-<WBR>9-<WBR>phenylxanthene
CAS Number:
Empirical Formula (Hill Notation):
 C19H13ClO
Molecular Weight:
 292.76
 Beilstein/REAXYS Number:
 237712
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More