• USA Home
  • Search Results

  • 9cisretinoicacid30044530003811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

9-cis-Retinoic acid

1 Products
Synonym:   9-cis-Tretinoin, Alitretinoin
Image of 9-<I>cis</I>-Retinoic acid
CAS Number:
Empirical Formula (Hill Notation):
 C20H28O2
Molecular Weight:
 300.44
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More