• USA Home
  • Search Results

  • 9decen1ol156271301922211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

9-Decen-1-ol

2 Products
Image of 9-Decen-1-ol
CAS Number:
Linear Formula:
 H2C=CH(CH2)8OH
Molecular Weight:
 156.27
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39020322
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More