• USA Home
  • Search Results

  • 9mesityl10methylacridiniumperchlorate4118867478397211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

9-Mesityl-10-methylacridinium perchlorate

1 Products
Synonym:   10-Methyl-9-(2,4,6-trimethylphenyl)acridinium perchlorate
Image of 9-<WBR>Mesityl-<WBR>10-<WBR>methylacridinium perchlorate
CAS Number:
Empirical Formula (Hill Notation):
 C23H22ClNO4
Molecular Weight:
 411.88
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More