• USA Home
  • Search Results

  • D6aminopenicillanicacid2162655116611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(+)-6-Aminopenicillanic acid

2 Products
Synonym:   (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-APA
Image of (+)<WBR>-<WBR>6-<WBR>Aminopenicillanic acid
CAS Number:
Empirical Formula (Hill Notation):
 C8H12N2O3S
Molecular Weight:
 216.26
 Beilstein/REAXYS Number:
 15080
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More