• USA Home
  • Search Results

  • Dborneol1542546443711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(+)-Borneol

2 Products
Synonym:   endo-(1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
Image of (+)-Borneol
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 2038056
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More