• USA Home
  • Search Results

  • Dnootkatone21833467450411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(+)-Nootkatone

3 Products
Image of (+)-Nootkatone
CAS Number:
Empirical Formula (Hill Notation):
 C15H22O
Molecular Weight:
 218.33
 Beilstein/REAXYS Number:
 4676969
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Melting Point
Show More